2-(Trifluoroacetyl)pyrrole 99%

Lieferant: Apollo Scientific
Achtung

PC3389-25G PC3389-5G PC3389-1G
APOSPC3389-25GEA 548 EUR
APOSPC3389-25G APOSPC3389-5G APOSPC3389-1G
2-(Trifluoroacetyl)pyrrole 99%
2-(Trifluoroacetyl)pyrrole
Formel: C₆H₄F₃NO
Molecular Weight: 163.100 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00141809
CAS-Nummer: 2557-70-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR