Trifluoroacetyl triflate 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Trifluoroacetyl Trifluoromethanesulphonate

PC3391-5G
APOSPC3391-5GEA 284 EUR
APOSPC3391-5G
Trifluoroacetyl triflate 96%
Trifluoroacetyl triflate
Formel: C₃F₆O₄S
Molecular Weight: 246,08 g/mol
Siedepunkt: 62…63 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011638
CAS-Nummer: 68602-57-3
UN: 2922
ADR: 8+6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR