Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphite, Phosphorous acid tris(1,1,1,3,3,3-hexafluoro-2-propyl) ester
PC3407-1G
APOSPC3407-1GEA
139
EUR
APOSPC3407-1G
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite
Formel:
C₉H₃F₁₈O₃P Molecular Weight: 532,07 g/mol Siedepunkt: 130 °C Dichte: 1,69 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192561 CAS-Nummer: 66470-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...