Methyl-4-hydroxy-6-(trifluoromethyl)thieno-[3,4-b]-pyridine-1-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

PC3409-250MG
APOSPC3409-250MGEA 46.4 EUR
APOSPC3409-250MG
Methyl-4-hydroxy-6-(trifluoromethyl)thieno-[3,4-b]-pyridine-1-carboxylate 97%
Methyl-4-hydroxy-6-(trifluoromethyl)thieno-[3,4-b]-pyridine-1-carboxylate
Formel: C₁₀H₆F₃NO₃S
Molecular Weight: 277,22 g/mol
Schmelzpunkt: 144…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173841
CAS-Nummer: 175203-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR