2-(Difluoromethoxy)phenol 98%

Lieferant: Apollo Scientific

PC3421-1G PC3421-250MG PC3421-500MG PC3421-5G
APOSPC3421-1GEA 104 EUR
APOSPC3421-1G APOSPC3421-250MG APOSPC3421-500MG APOSPC3421-5G
2-(Difluoromethoxy)phenol 98%
Formel: C₇H₆F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00236234
CAS-Nummer: 53104-96-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR