3'-Fluor-4'-(trifluormethyl)acetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanone

PC3426-25G PC3426-5G PC3426-1G
APOSPC3426-25GEA 342 EUR
APOSPC3426-25G APOSPC3426-5G APOSPC3426-1G
3'-Fluor-4'-(trifluormethyl)acetophenon 98%
3'-Fluor-4'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00236273
CAS-Nummer: 237761-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR