Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 1,4-dihydro-8-fluor-4-oxoquinoline-3-carboxylat

PC3427-1G
APOSPC3427-1GEA 21.9 EUR
APOSPC3427-1G
Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat
Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀FNO₃
Molecular Weight: 235.211 g/mol
Schmelzpunkt: 217…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052242
CAS-Nummer: 63010-69-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR