2-(Difluormethoxy)benzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC3432-250MG
APOSPC3432-250MGEA 14.7 EUR
APOSPC3432-250MG
2-(Difluormethoxy)benzoesäure 98%
2-(Difluormethoxy)benzoesäure
Formel: C₈H₆F₂O₃
Molecular Weight: 188.13 g/mol
Schmelzpunkt: 98…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236223
CAS-Nummer: 97914-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR