2-(Trifluormethoxy)benzolsulfonamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Trifluormethoxy)benzolsulfonsäureamid

PC3435-1G PC3435-5G PC3435-25G
APOSPC3435-1GEA 14.7 EUR
APOSPC3435-1G APOSPC3435-5G APOSPC3435-25G
2-(Trifluormethoxy)benzolsulfonamid
2-(Trifluormethoxy)benzolsulfonamid
Formel: C₇H₆F₃NO₃S
Molecular Weight: 241.19 g/mol
Schmelzpunkt: 187…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320751
CAS-Nummer: 37526-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR