2-(Ethoxycarbonyl)-5-fluorphenylboronsäure

Lieferant: Apollo Scientific

Synonyme: 2-Ethoxycabonyl-5-fluorophenylboronic acid, (2-(Ethoxycarbonyl)-5-fluorophenyl)boronic acid

PC3438-1G
APOSPC3438-1GEA 176 EUR
APOSPC3438-1G
2-(Ethoxycarbonyl)-5-fluorphenylboronsäure
2-(Ethoxycarbonyl)-5-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211,9800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878342
CAS-Nummer: 957062-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR