2-Bromo-6-fluoro-3-methoxybenzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: (2-Bromo-6-fluoro-3-methoxyphenyl)boronic acid

PC3439-100G PC3439-25G PC3439-5G
APOSPC3439-100GEA 564 EUR
APOSPC3439-100G APOSPC3439-25G APOSPC3439-5G
2-Bromo-6-fluoro-3-methoxybenzeneboronic acid
2-Bromo-6-fluoro-3-methoxybenzeneboronic acid
Formel: C₇H₇BBrFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08276828
CAS-Nummer: 957062-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR