3-Brom-N,N-dipropyl-5-(trifluormethyl)benzolsulfonamid

Lieferant: Apollo Scientific

PC3448-1G
APOSPC3448-1GEA 263 EUR
APOSPC3448-1G
3-Brom-N,N-dipropyl-5-(trifluormethyl)benzolsulfonamid
3-Brom-N,N-dipropyl-5-(trifluormethyl)benzolsulfonamid
Formel: C₁₃H₁₇BrF₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09878371
CAS-Nummer: 957062-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR