2-(4-Fluorobenzenesulphonyl)acetamide oxime 97%

Lieferant: Apollo Scientific
Achtung

PC3449-1G
APOSPC3449-1GEA 88.6 EUR
APOSPC3449-1G
2-(4-Fluorobenzenesulphonyl)acetamide oxime 97%
2-(4-Fluorobenzenesulphonyl)acetamide oxime
Formel: C₈H₉FN₂O₃S
Molecular Weight: 232,23 g/mol
Schmelzpunkt: 180…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00205110
CAS-Nummer: 175203-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR