1-(4-Fluor-2-nitrophenyl)piperidin

Lieferant: Apollo Scientific
Achtung

PC3451-1G
APOSPC3451-1GEA 0 EUR
APOSPC3451-1G
1-(4-Fluor-2-nitrophenyl)piperidin
1-(4-Fluor-2-nitrophenyl)piperidin
Formel: C₁₁H₁₃FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00798370
CAS-Nummer: 719-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR