6-Fluor-o-anisaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Fluor-2-anisaldehyd, 2-Fluor-6-methoxybenzaldehyd
PC3458-1G
PC3458-25G
PC3458-5G
APOSPC3458-1GEA
21.9
EUR
APOSPC3458-1G
APOSPC3458-25G
APOSPC3458-5G
6-Fluor-o-anisaldehyd 98%
6-Fluor-o-anisaldehyd
Formel:
C₈H₇FO₂ Molecular Weight: 154.14 g/mol Schmelzpunkt: 59…63 °C Flammpunkt: 110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01090998 CAS-Nummer: 146137-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...