1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(5-(Trifluoromethyl)pyridin-2-yl)piperazine

PC3465-5G
APOSPC3465-5GEA 21.9 EUR
APOSPC3465-5G
1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine 97%
1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine
Formel: C₁₀H₁₂F₃N₃
Molecular Weight: 231,222 g/mol
Siedepunkt: 122 °C/2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00114708
CAS-Nummer: 132834-58-3
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR