2-Fluorobenzal bromide

Lieferant: Apollo Scientific
Gefahr

PC3471-1G PC3471-5G
APOSPC3471-1GEA 0 EUR
APOSPC3471-1G APOSPC3471-5G
2-Fluorobenzal bromide
2-Fluorobenzal bromide
Formel: C₇H₅Br₂F
Lagertemperatur: Raumtemperatur
MDL: MFCD00045061
CAS-Nummer: 220141-76-4
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR