4-Fluorbenzamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluorbenzoesäureamid

PC3476-100G
APOSPC3476-100GEA 31.9 EUR
APOSPC3476-100G
4-Fluorbenzamid
4-Fluorbenzamid
Formel: C₇H₆FNO
Molecular Weight: 139,13 g/mol
Schmelzpunkt: 154…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007992
CAS-Nummer: 824-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR