2,2-Bis(4-isocyanatophenyl)hexafluoropropane 98%

Lieferant: Apollo Scientific

Synonyme: 4,4'-(Hexafluoroisopropylidene)bis(isocyanatobenzene)

PC3481-1G
APOSPC3481-1GEA 270 EUR
APOSPC3481-1G
2,2-Bis(4-isocyanatophenyl)hexafluoropropane 98%
2,2-Bis(4-isocyanatophenyl)hexafluoropropane

  • May form small amount of sediment.

Formel: C₁₇H₈F₆N₂O₂
Molecular Weight: 386.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00142759
CAS-Nummer: 10224-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR