4-Fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

PC3482-25G PC3482-100G PC3482-500G
APOSPC3482-25GEA 0 EUR
APOSPC3482-25G APOSPC3482-100G APOSPC3482-500G
4-Fluorphenylboronsäure 97%
4-Fluorphenylboronsäure

Contains variable amounts of the anhydride GLC reagent for diols see: J. Chromatogr.,158,33,1978

Formel: C₆H₆BFO₂
Schmelzpunkt: 262…265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039136
CAS-Nummer: 1765-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR