1-(3-Fluorophenyl)propylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Amino-1-(3-fluorophenyl)propane, α-Ethyl-3-fluorobenzylamine

PC3484-250MG
APOSPC3484-250MGEA 158 EUR
APOSPC3484-250MG
1-(3-Fluorophenyl)propylamine
1-(3-Fluorophenyl)propylamine
Formel: C₉H₁₂FN
Flammpunkt: 85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784284
CAS-Nummer: 473732-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR