[(4-Fluorophenyl)sulphonyl]acetonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3487M-1G
APOSPC3487M-1GEA
109
EUR
APOSPC3487M-1G
[(4-Fluorophenyl)sulphonyl]acetonitrile 97%
[(4-Fluorophenyl)sulphonyl]acetonitrile
Formel:
C₈H₆FNO₂S Schmelzpunkt: 87…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00179339 CAS-Nummer: 32083-66-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...