2-Brom-N-ethyl-4-(trifluormethoxy)benzolsulfonamid
Lieferant: Apollo Scientific
PC3492-1G
APOSPC3492-1GEA
357
EUR
APOSPC3492-1G
2-Brom-N-ethyl-4-(trifluormethoxy)benzolsulfonamid
2-Brom-N-ethyl-4-(trifluormethoxy)benzolsulfonamid
Formel:
C₉H₉BrF₃NO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878394 CAS-Nummer: 957062-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...