2-Brom-N-butyl-4-(trifluormethoxy)benzolsulfonamid

Lieferant: Apollo Scientific

PC3493-1G
APOSPC3493-1GEA 248 EUR
APOSPC3493-1G
2-Brom-N-butyl-4-(trifluormethoxy)benzolsulfonamid
2-Brom-N-butyl-4-(trifluormethoxy)benzolsulfonamid
Formel: C₁₁H₁₃BrF₃NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD09878395
CAS-Nummer: 957062-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR