2-Brom-N-cyclopropyl-4-(trifluormethoxy)benzolsulfonamid

Lieferant: Apollo Scientific

PC3494-1G
APOSPC3494-1GEA 357 EUR
APOSPC3494-1G
2-Brom-N-cyclopropyl-4-(trifluormethoxy)benzolsulfonamid
Formel: C₁₀H₉BrF₃NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD09878380
CAS-Nummer: 957062-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR