1-[2-Fluoro-4-(methylsulphonyl)phenyl]-3-methylpiperazine

Lieferant: Apollo Scientific
Achtung

PC3507-1G
APOSPC3507-1GEA 371 EUR
APOSPC3507-1G
1-[2-Fluoro-4-(methylsulphonyl)phenyl]-3-methylpiperazine
1-[2-Fluoro-4-(methylsulphonyl)phenyl]-3-methylpiperazine
Formel: C₁₂H₁₇FN₂O₂S
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692476
CAS-Nummer: 914637-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR