2-[4-[2-Fluoro-4-(methylsulphonyl)phenyl]piperazin-1-yl]ethanol

Lieferant: Apollo Scientific

PC3508-1G
APOSPC3508-1GEA 270 EUR
APOSPC3508-1G
2-[4-[2-Fluoro-4-(methylsulphonyl)phenyl]piperazin-1-yl]ethanol
2-[4-[2-Fluoro-4-(methylsulphonyl)phenyl]piperazin-1-yl]ethanol
Formel: C₁₃H₁₉FN₂O₃S
Schmelzpunkt: 132…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03849563
CAS-Nummer: 887453-53-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR