1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-4-carboxamide

Lieferant: Apollo Scientific

PC3509-1G
APOSPC3509-1GEA 335 EUR
APOSPC3509-1G
1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-4-carboxamide
1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-4-carboxamide
Formel: C₁₃H₁₇FN₂O₃S
Schmelzpunkt: 226…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692427
CAS-Nummer: 914637-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR