1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperidine-4-carboxamide

Lieferant: Apollo Scientific

PC3511-500MG
APOSPC3511-500MGEA 285 EUR
APOSPC3511-500MG
1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperidine-4-carboxamide
1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperidine-4-carboxamide
Formel: C₁₃H₁₇FN₂O₃S
Molecular Weight: 300.35 g/mol
Schmelzpunkt: 160…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692469
CAS-Nummer: 914637-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR