1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC3513-500MG
APOSPC3513-500MGEA 335 EUR
APOSPC3513-500MG
1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-3-carboxylic acid
1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-3-carboxylic acid
Formel: C₁₃H₁₆FNO₄S
Molecular Weight: 301.34 g/mol
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692426
CAS-Nummer: 914637-67-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR