1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-4-carboxylic acid

Lieferant: Apollo Scientific

PC3514-1G
APOSPC3514-1GEA 383 EUR
APOSPC3514-1G
1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-4-carboxylic acid
1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperidine-4-carboxylic acid
Formel: C₁₃H₁₆FNO₄S
Molecular Weight: 301,334 g/mol
Schmelzpunkt: 189…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692429
CAS-Nummer: 914637-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR