1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperidine-4-carboxylic acid

Lieferant: Apollo Scientific

PC3517-500MG
APOSPC3517-500MGEA 303 EUR
APOSPC3517-500MG
1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperidine-4-carboxylic acid
1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperidine-4-carboxylic acid
Formel: C₁₃H₁₆FNO₄S
Molecular Weight: 301,334 g/mol
Schmelzpunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692468
CAS-Nummer: 914637-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR