8-(2-Fluoro-4-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
Lieferant: Apollo Scientific
PC3518-1G
APOSPC3518-1GEA
270
EUR
APOSPC3518-1G
8-(2-Fluoro-4-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
8-(2-Fluoro-4-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
Formel:
C₁₃H₁₅FN₂O₄ Schmelzpunkt: 123…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08692382 CAS-Nummer: 252336-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...