8-(4-Fluoro-2-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
Lieferant: Apollo Scientific
PC3519-1G
APOSPC3519-1GEA
183
EUR
APOSPC3519-1G
8-(4-Fluoro-2-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
8-(4-Fluoro-2-nitrophenyl)-1,4-dioxa-8-azaspiro[4.5]decane
Formel:
C₁₃H₁₅FN₂O₄ Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08692464 CAS-Nummer: 914637-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...