1-(4-Fluoro-2-nitrophenyl)homopiperazine hydrochloride

Lieferant: Apollo Scientific

PC3523-500MG
APOSPC3523-500MGEA 270 EUR
APOSPC3523-500MG
1-(4-Fluoro-2-nitrophenyl)homopiperazine hydrochloride
1-(4-Fluoro-2-nitrophenyl)homopiperazine hydrochloride
Formel: C₁₁H₁₅ClFN₃O₂
Schmelzpunkt: 179…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692456
CAS-Nummer: 1185298-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR