1-(4-Fluoro-2-nitrophenyl)-3-methylpiperazine hydrochloride

Lieferant: Apollo Scientific

PC3525-1G
APOSPC3525-1GEA 187 EUR
APOSPC3525-1G
1-(4-Fluoro-2-nitrophenyl)-3-methylpiperazine hydrochloride
1-(4-Fluoro-2-nitrophenyl)-3-methylpiperazine hydrochloride
Formel: C₁₁H₁₅ClFN₃O₂
Schmelzpunkt: 208…209 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692452
CAS-Nummer: 1185298-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR