1-(4-Fluoro-2-nitrophenyl)-2-methylpiperidine hydrochloride

Lieferant: Apollo Scientific

PC3526-1G
APOSPC3526-1GEA 183 EUR
APOSPC3526-1G
1-(4-Fluoro-2-nitrophenyl)-2-methylpiperidine hydrochloride
1-(4-Fluoro-2-nitrophenyl)-2-methylpiperidine hydrochloride
Formel: C₁₂H₁₆ClFN₂O₂
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692444
CAS-Nummer: 1185302-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR