2-Fluor-5-(methylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Fluoro-5-(methylcarbamoyl)phenyl)boronic acid, 2-Fluoro-5-(methylcarbamoyl)benzeneboronic acid

PC3527-1G
APOSPC3527-1GEA 68.3 EUR
APOSPC3527-1G
2-Fluor-5-(methylcarbamoyl)phenylboronsäure 98%
2-Fluor-5-(methylcarbamoyl)phenylboronsäure
Formel: C₈H₉BFNO₃
Molecular Weight: 196.97 g/mol
Schmelzpunkt: 174…176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436028
CAS-Nummer: 874289-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR