5-(Ethylcarbamoyl)-2-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-(Ethylcarbamoyl)-2-fluorophenyl)boronic acid

PC3528-1G
APOSPC3528-1GEA 92.8 EUR
APOSPC3528-1G
5-(Ethylcarbamoyl)-2-fluorphenylboronsäure 97%
5-(Ethylcarbamoyl)-2-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Molecular Weight: 211,0000 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436027
CAS-Nummer: 874289-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR