2-Fluor-5-(propylcarbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (2-Fluoro-5-(propylcarbamoyl)phenyl)boronic acid

PC3529-1G
APOSPC3529-1GEA 248 EUR
APOSPC3529-1G
2-Fluor-5-(propylcarbamoyl)phenylboronsäure 97%
2-Fluor-5-(propylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BFNO₃
Molecular Weight: 225,0300 g/mol
Schmelzpunkt: 147…149 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436029
CAS-Nummer: 874289-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR