3-Fluor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-4-(pyrolidine-1-carbonyl)phenylboronic acid, (3-Fluoro-4-(pyrrolidine-1-carbonyl)phenyl)boronic acid

PC3535-1G
APOSPC3535-1GEA 277 EUR
APOSPC3535-1G
3-Fluor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure 98%
3-Fluor-4-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₃
Molecular Weight: 237,0400 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436024
CAS-Nummer: 874289-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR