2-Fluor-(4-methoxycarbonyl)phenylboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-4-methoxycarbonylphenylboronic acid
PC3537-1G
PC3537-250MG
PC3537-5G
APOSPC3537-1GEA
24.8
EUR
APOSPC3537-1G
APOSPC3537-250MG
APOSPC3537-5G
2-Fluor-(4-methoxycarbonyl)phenylboronsäure 96%
2-Fluor-(4-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BFO₄ Schmelzpunkt: 173…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436034 CAS-Nummer: 603122-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...