3-Fluor-4-(methoxy(methyl)carbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

PC3556-1G PC3556-5G
APOSPC3556-1GEA 163 EUR
APOSPC3556-1G APOSPC3556-5G
3-Fluor-4-(methoxy(methyl)carbamoyl)phenylboronsäure 98%
3-Fluor-4-(methoxy(methyl)carbamoyl)phenylboronsäure
Formel: C₉H₁₁BFNO₄
Schmelzpunkt: 154…156 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689477
CAS-Nummer: 913835-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR