2-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (2-Fluoro-5-(piperidine-1-carbonyl)phenyl)boronic acid, 2-Fluoro-5-(1-piperidinylcarbonyl)benzeneboronic acid

PC3562-1G
APOSPC3562-1GEA 248 EUR
APOSPC3562-1G
2-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure 97%
2-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BFNO₃
Molecular Weight: 251,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436068
CAS-Nummer: 874289-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR