4-(Diethylcarbamoyl)-3-fluorobenzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Diethylcarbamoyl)-3-fluorophenylboronic acid, (4-(Diethylcarbamoyl)-3-fluorophenyl)boronic acid

PC3568-1G
APOSPC3568-1GEA 176 EUR
APOSPC3568-1G
4-(Diethylcarbamoyl)-3-fluorobenzeneboronic acid 98%
4-(Diethylcarbamoyl)-3-fluorobenzeneboronic acid
Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239,051 g/mol
Schmelzpunkt: 158…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436057
CAS-Nummer: 874289-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR