3-Bromo-5-fluorobenzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-fluorophenylboronic acid, (3-Bromo-5-fluorophenyl)boronic acid

PC3569-1G
APOSPC3569-1GEA 21.9 EUR
APOSPC3569-1G
3-Bromo-5-fluorobenzeneboronic acid 95%
3-Bromo-5-fluorobenzeneboronic acid
Formel: C₆H₅BBrFO₂
Molecular Weight: 218,82 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06411356
CAS-Nummer: 849062-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR