5-Benzylcarbamoyl-2-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (5-(Benzylcarbamoyl)-2-fluorophenyl)boronic acid

PC3582-1G
APOSPC3582-1GEA 248 EUR
APOSPC3582-1G
5-Benzylcarbamoyl-2-fluorphenylboronsäure 97%
5-Benzylcarbamoyl-2-fluorphenylboronsäure
Formel: C₁₄H₁₃BFNO₃
Schmelzpunkt: 224…226 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436031
CAS-Nummer: 874289-53-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR