7-Fluoroisatin-3-oxime 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Dihydro-7-fluoro-3-(hydroxyimino)-2H-indol-2-one, 7-Fluoro-1H-indole-2,3-dione 3-oxime
PC3583-1G
PC3583-25G
PC3583-5G
APOSPC3583-1GEA
21.9
EUR
APOSPC3583-1G
APOSPC3583-25G
APOSPC3583-5G
7-Fluoroisatin-3-oxime 98%
7-Fluoroisatin-3-oxime
Formel:
C₈H₅FN₂O₂ Molecular Weight: 180.136 g/mol Schmelzpunkt: 207…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02951896 CAS-Nummer: 143884-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...