7-Fluoroisatin-3-oxime 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Dihydro-7-fluoro-3-(hydroxyimino)-2H-indol-2-one, 7-Fluoro-1H-indole-2,3-dione 3-oxime

PC3583-1G PC3583-25G PC3583-5G
APOSPC3583-1GEA 21.9 EUR
APOSPC3583-1G APOSPC3583-25G APOSPC3583-5G
7-Fluoroisatin-3-oxime 98%
7-Fluoroisatin-3-oxime
Formel: C₈H₅FN₂O₂
Molecular Weight: 180.136 g/mol
Schmelzpunkt: 207…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02951896
CAS-Nummer: 143884-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR