2-(Trifluormethylthio)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC3586-10G
PC3586-5G
APOSPC3586-10GEA
0
EUR
APOSPC3586-10G
APOSPC3586-5G
2-(Trifluormethylthio)benzylbromid 98%
2-(Trifluormethylthio)benzylbromid
Formel:
C₈H₆BrF₃S Siedepunkt: 71 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236345 CAS-Nummer: 238403-52-6 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...